Gene Transcripts

Gene Symbol
Srpx2
Entrez Gene ID
317181
Gene Name
sushi-repeat-containing protein, X-linked 2
Gene Aliases
RGD1562444
Chromosome Location
Chr. X: 104734368 - 104760658 on Build Rnor_6.0
UniGene ID
Rn.33831
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108243.2 NP_001101713.1 7, 8 922
GenBank mRNA BC168975.1 - 7, 8 922

View Full Product Details