Gene Transcripts

Gene Symbol
Rasgef1c
Entrez Gene ID
360519
Gene Name
RasGEF domain family, member 1C
Gene Aliases
Chromosome Location
Chr. 10: 35392762 - 35459267 on Build Rnor_6.0
UniGene ID
Rn.46134
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108273.1 NP_001101743.1 11 1290

View Full Product Details