Gene Transcripts
- Gene Symbol
- SNX3
- Entrez Gene ID
- 8724
- Gene Name
- sorting nexin 3
- Gene Aliases
- Grd19, MCOPS8, SDP3
- Chromosome Location
- Chr. 6: 108211217 - 108261260 on Build GRCh38
- UniGene ID
- Hs.12102
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300928.1 |
NP_001287857.1
|
1, 2 |
508 |
|
NM_003795.5 |
NP_003786.1
|
1, 2 |
508 |
|
GenBank mRNA |
AF034546.1 |
- |
1, 2 |
212 |
|
AF062483.1 |
- |
1, 2 |
188 |
|
AK289476.1 |
- |
1, 2 |
299 |
|
BC008444.1 |
- |
1, 2 |
266 |
4102735 |
BC014580.1 |
- |
1, 2 |
290 |
3932208 |
BC015179.1 |
- |
1, 2 |
272 |
3905515 |
BC016863.2 |
- |
1, 2 |
522 |
3846511 |
BG497532.1 |
- |
1, 2 |
506 |
|
BT007114.1 |
- |
1, 2 |
169 |
|
CR456898.1 |
- |
1, 2 |
169 |
|
HY156437.1 |
- |
1, 2 |
346 |
|
View Full Product Details