Gene Transcripts

Gene Symbol
Lypd1
Entrez Gene ID
360838
Gene Name
Ly6/Plaur domain containing 1
Gene Aliases
Lypdc1
Chromosome Location
Chr. 13: 42219884 - 42263024 on Build Rnor_6.0
UniGene ID
Rn.231867
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007727.1 NP_001007728.1 3 572
GenBank mRNA BC082032.1 - 3 572

View Full Product Details