Gene Transcripts

Gene Symbol
Ccdc25
Entrez Gene ID
361059
Gene Name
coiled-coil domain containing 25
Gene Aliases
RGD1307689
Chromosome Location
Chr. 15: 42522108 - 42555318 on Build Rnor_6.0
UniGene ID
Rn.136803
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108382.1 NP_001101852.1 7 491
XM_006252132.2 XP_006252194.1 7 490

View Full Product Details