Gene Transcripts

Gene Symbol
Zic5
Entrez Gene ID
361095
Gene Name
Zic family member 5
Gene Aliases
Chromosome Location
Chr. 15: 108891992 - 108898703 on Build Rnor_6.0
UniGene ID
Rn.198356
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108391.1 NP_001101861.1 3 1194

View Full Product Details