Gene Transcripts

Gene Symbol
Rras
Entrez Gene ID
361568
Gene Name
related RAS viral (r-ras) oncogene homolog
Gene Aliases
p23
Chromosome Location
Chr. 1: 101012822 - 101016720 on Build Rnor_6.0
UniGene ID
Rn.71224
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108481.1 NP_001101951.1 6 905
GenBank mRNA FQ211166.1 - 6 921

View Full Product Details