Gene Transcripts
- Gene Symbol
- BRSK2
- Entrez Gene ID
- 9024
- Gene Name
- BR serine/threonine kinase 2
- Gene Aliases
- C11orf7, PEN11B, SAD1, SADA, STK29
- Chromosome Location
- Chr. 11: 1389899 - 1462689 on Build GRCh38
- UniGene ID
- Hs.170819
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256627.1 |
NP_001243556.1
|
18, 19 |
2323 |
|
NM_001256630.1 |
NP_001243559.1
|
18, 19 |
2328 |
|
NM_003957.3 |
NP_003948.2
|
18, 19 |
2323 |
|
XM_005253218.3 |
XP_005253275.1
|
18, 19 |
2035 |
|
XM_006718374.2 |
XP_006718437.1
|
19, 20 |
2101 |
|
XM_006718376.2 |
XP_006718439.1
|
19, 20 |
2013 |
|
XM_011520461.2 |
XP_011518763.1
|
18, 19 |
1947 |
|
XM_011520462.2 |
XP_011518764.1
|
18, 19 |
1947 |
|
XM_017018532.1 |
XP_016874021.1
|
19, 20 |
2379 |
|
XM_017018533.1 |
XP_016874022.1
|
19, 20 |
2467 |
|
XM_017018534.1 |
XP_016874023.1
|
19, 20 |
2379 |
|
XM_017018535.1 |
XP_016874024.1
|
18, 19 |
2313 |
|
XM_017018536.1 |
XP_016874025.1
|
18, 19 |
2401 |
|
XM_017018537.1 |
XP_016874026.1
|
19, 20 |
2379 |
|
XM_017018538.1 |
XP_016874027.1
|
14, 15 |
1924 |
|
GenBank mRNA |
AF020089.1 |
- |
5, 6 |
551 |
|
AF533876.1 |
- |
18, 19 |
2321 |
|
AF533879.1 |
- |
18, 19 |
2321 |
|
AK131534.1 |
- |
18, 19 |
2328 |
|
AY166857.1 |
- |
18, 19 |
2323 |
|
View Full Product Details