Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001044253.1 | NP_001037718.1 | 5 | 1062 | |
XM_006243271.2 | XP_006243333.1 | 5 | 1999 | ||
XM_008766324.1 | XP_008764546.1 | 6 | 1718 | ||
XM_008766325.1 | XP_008764547.1 | 6 | 1690 | ||
XM_008766326.1 | XP_008764548.1 | 6 | 1649 | ||
XM_008766327.1 | XP_008764549.1 | 6 | 1621 | ||
GenBank mRNA | DQ489562.1 | - | 5 | 1062 |