Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005445.3 | NP_005436.1 | 15 | 1613 | |
GenBank mRNA | AF020043.1 | - | 15 | 1578 | |
AF067163.1 | - | 3 | 260 | ||
AK289771.1 | - | 15 | 1609 | ||
AK315845.1 | - | 15 | 1612 | ||
BC047324.1 | - | 15 | 1508 | 5493263 |