Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004719.2 | NP_004710.2 | 12 | 3893 | |
XM_005269230.2 | XP_005269287.2 | 12 | 3783 | ||
XM_011538984.2 | XP_011537286.1 | 13 | 5412 | ||
XM_011538985.1 | XP_011537287.1 | 12 | 3727 | ||
XM_017020217.1 | XP_016875706.1 | 13 | 3771 | ||
XM_017020218.1 | XP_016875707.1 | 13 | 3771 | ||
XM_017020219.1 | XP_016875708.1 | 13 | 3970 | ||
XM_017020220.1 | XP_016875709.1 | 13 | 3771 | ||
XM_017020221.1 | XP_016875710.1 | 11 | 4100 | ||
XM_017020222.1 | XP_016875711.1 | 10 | 4070 | ||
XM_017020223.1 | XP_016875712.1 | 13 | 3993 | ||
GenBank mRNA | AF030234.1 | - | 12 | 3841 | |
AK314093.1 | - | 6 | 3156 | ||
Y11251.1 | - | 2 | 3871 |