Gene Transcripts

Gene Symbol
Vom1r25
Entrez Gene ID
494270
Gene Name
vomeronasal 1 receptor 25
Gene Aliases
V1rm2
Chromosome Location
Chr. 8: 3540361 - 3541275 on Build Rnor_6.0
UniGene ID
Rn.129441
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008921.1 NP_001008921.1 1 161
GenBank mRNA AY510371.1 - 1 161

View Full Product Details