Gene Transcripts

Gene Symbol
Trim30
Entrez Gene ID
499223
Gene Name
tripartite motif-containing 30
Gene Aliases
RGD1563970
Chromosome Location
Chr. 1: 169521655 - 169540237 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229917.2 XP_006229979.1 3 758

View Full Product Details