Gene Transcripts
- Gene Symbol
- Sprr1a
- Entrez Gene ID
- 499660
- Gene Name
- small proline-rich protein 1A
- Gene Aliases
- Spr, Spr1a, Sprr, Sprr1, Sprr1al, Sprr1b
- Chromosome Location
- Chr. 2: 192669143 - 192671059 on Build Rnor_6.0
- UniGene ID
- Rn.14720
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021864.1 |
NP_068636.1
|
2 |
151 |
|
View Full Product Details