Gene Transcripts

Gene Symbol
Pex1
Entrez Gene ID
500006
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
RGD1559939
Chromosome Location
Chr. 4: 27659376 - 27698037 on Build Rnor_6.0
UniGene ID
Rn.162727
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109220.1 NP_001102690.1 12, 13 2073
XM_006236054.1 XP_006236116.1 12, 13 2107

View Full Product Details