Gene Transcripts
- Gene Symbol
- HS6ST1
- Entrez Gene ID
- 9394
- Gene Name
- heparan sulfate 6-O-sulfotransferase 1
- Gene Aliases
- HH15, HS6ST
- Chromosome Location
- Chr. 2: 128265480 - 128318597 on Build GRCh38
- UniGene ID
- Hs.512841
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004807.2 |
NP_004798.3
|
1, 2 |
568 |
|
GenBank mRNA |
AB006179.1 |
- |
1, 2 |
642 |
|
AK293724.1 |
- |
1, 2 |
25 |
|
BC096239.1 |
- |
1, 2 |
544 |
40005544 |
BC096240.1 |
- |
1, 2 |
544 |
40005547 |
BC099638.3 |
- |
1, 2 |
544 |
40005546 |
BC099639.1 |
- |
1, 2 |
544 |
40005548 |
BP219645.1 |
- |
1, 2 |
454 |
|
CV569969.1 |
- |
2, 3 |
273 |
|
DR005030.1 |
- |
1, 2 |
574 |
|
View Full Product Details