Gene Transcripts

Gene Symbol
Prss44
Entrez Gene ID
501060
Gene Name
protease, serine, 44
Gene Aliases
RGD1560940, Tessp4
Chromosome Location
Chr. 8: 119083900 - 119097707 on Build Rnor_6.0
UniGene ID
Rn.126314
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109298.1 NP_001102768.1 1 305
XM_008766647.1 XP_008764869.1 1 305
GenBank mRNA BC168977.1 - 1 284

View Full Product Details