Gene Transcripts

Gene Symbol
LOC102546630
Entrez Gene ID
102546630
Gene Name
protein gar2-like
Gene Aliases
Chromosome Location
Chr. X: 63998813 - 63999664 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257041.2 XP_006257103.1 1 257

Gene Symbol
LOC102551035
Entrez Gene ID
102551035
Gene Name
serine/arginine repetitive matrix protein 4-like
Gene Aliases
Chromosome Location
Chr. X: 65335316 - 65336069 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257045.2 XP_006257107.1 1 171

Gene Symbol
LOC103694487
Entrez Gene ID
103694487
Gene Name
protein gar2-like
Gene Aliases
Chromosome Location
Chr. X: 63897678 - 63898354 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773293.1 XP_008771515.1 1 193

View Full Product Details