Gene Transcripts

Gene Symbol
Rhox10
Entrez Gene ID
652926
Gene Name
reproductive homeobox 10
Gene Aliases
Chromosome Location
Chr. X: 124207017 - 124211455 on Build Rnor_6.0
UniGene ID
Rn.144547
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037581.1 NP_001032670.1 2, 3 413
GenBank mRNA DQ058657.1 - 2, 3 413

View Full Product Details