Gene Transcripts

Gene Symbol
Paqr6
Entrez Gene ID
681021
Gene Name
progestin and adipoQ receptor family member VI
Gene Aliases
Chromosome Location
Chr. 2: 187736765 - 187741696 on Build Rnor_6.0
UniGene ID
Rn.204669
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191077.1 NP_001178006.1 4 338
XM_006232755.2 XP_006232817.1 4 1340
XM_008761210.1 XP_008759432.1 4 1340
XM_008761212.1 XP_008759434.1 4 445

View Full Product Details