Gene Transcripts

Gene Symbol
Asb14
Entrez Gene ID
680076
Gene Name
ankyrin repeat and SOCS box-containing 14
Gene Aliases
Chromosome Location
Chr. 16: 2537214 - 2556905 on Build Rnor_6.0
UniGene ID
Rn.19511
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170749.1 NP_001164220.1 8 1335
XM_008771008.1 XP_008769230.1 9 1439
XM_008771009.1 XP_008769231.1 3 749

View Full Product Details