Gene Transcripts

Gene Symbol
Fam19a3
Entrez Gene ID
691123
Gene Name
family with sequence similarity 19 (chemokine (C-C motif)-like), member A3
Gene Aliases
Chromosome Location
Chr. 2: 207249000 - 207258898 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006233103.2 XP_006233165.1 2 638
XM_008761434.1 XP_008759656.1 2 638

View Full Product Details