Gene Transcripts

Gene Symbol
Kncn
Entrez Gene ID
690105
Gene Name
kinocilin
Gene Aliases
Chromosome Location
Chr. 5: 134736603 - 134744683 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729513.4 XP_002729559.2 3 659

View Full Product Details