Gene Transcripts

Gene Symbol
Ugt2b17
Entrez Gene ID
266685
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide B15
Gene Aliases
Rlug38, UDPGT 2B5, Udpgt, Udpgtr-3, Ugt2b3, Ugt2b5
Chromosome Location
Chr. 14: 22374877 - 22486291 on Build Rnor_6.0
UniGene ID
Rn.24945
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770000.1 XP_008768222.1 5 1295

Gene Symbol
Ugt2a3
Entrez Gene ID
289533
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide A3
Gene Aliases
RGD1308444
Chromosome Location
Chr. 14: 22251507 - 22267899 on Build Rnor_6.0
UniGene ID
Rn.124736
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135869.1 NP_001129341.1 1264

View Full Product Details