Gene Transcripts

Gene Symbol
LOC690183
Entrez Gene ID
690183
Gene Name
similar to DnaJ homolog subfamily B member 6 (Heat shock protein J2) (HSJ-2) (MRJ) (mDj4)
Gene Aliases
Chromosome Location
Chr. 1: 223548728 - 223550015 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001073579.4 XP_001073579.1 1 941

Gene Symbol
Dnajb6
Entrez Gene ID
362293
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 6
Gene Aliases
Chromosome Location
Chr. 4: 2711339 - 2774969 on Build Rnor_6.0
UniGene ID
Rn.129050
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA FQ214601.1 - 8 949

View Full Product Details