Gene Transcripts

Gene Symbol
Magohb
Entrez Gene ID
690303
Gene Name
mago-nashi homolog B (Drosophila)
Gene Aliases
Chromosome Location
Chr. 4: 165514582 - 165522366 on Build Rnor_6.0
UniGene ID
Rn.220911
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191105.1 NP_001178034.1 4 328
XM_008763423.1 XP_008761645.1 4 1604
XM_008763424.1 XP_008761646.1 5 473

View Full Product Details