Gene Transcripts

Gene Symbol
Tmem71
Entrez Gene ID
690657
Gene Name
transmembrane protein 71
Gene Aliases
Chromosome Location
Chr. 7: 107264235 - 107392808 on Build Rnor_6.0
UniGene ID
Rn.32320
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271215.1 NP_001258144.1 7 958

View Full Product Details