Gene Transcripts

Gene Symbol
Vom2r22
Entrez Gene ID
690758
Gene Name
vomeronasal 2 receptor, 22
Gene Aliases
Chromosome Location
Chr. 8: 4349354 - 4376039 on Build Rnor_6.0
UniGene ID
Rn.198478
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099656.1 NP_001093126.1 2029
XM_003750425.1 XP_003750473.1 6 2029

Gene Symbol
LOC100911044
Entrez Gene ID
100911044
Gene Name
vomeronasal type-2 receptor 26-like
Gene Aliases
Chromosome Location
Chr. 1: 62948554 - 62979947 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748732.1 XP_003748780.1 6 2044

View Full Product Details