Gene Transcripts

Gene Symbol
RRAS
Entrez Gene ID
6237
Gene Name
related RAS viral (r-ras) oncogene homolog
Gene Aliases
Chromosome Location
Chr. 19: 49635292 - 49640143 on Build GRCh38
UniGene ID
Hs.515536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006270.4 NP_006261.1 6 818
GenBank mRNA BC016286.1 - 6 797
BC016318.1 - 6 822 4073829

View Full Product Details