Gene Transcripts

Gene Symbol
HIST1H3I
Entrez Gene ID
8354
Gene Name
histone cluster 1, H3i
Gene Aliases
H3.f, H3/f, H3FF
Chromosome Location
Chr. 6: 27871845 - 27872321 on Build GRCh38
UniGene ID
Hs.132854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003533.2 NP_003524.1 1 417
GenBank mRNA BC066884.1 - 1 417
BC067493.1 - 1 417 7002079
BC069305.1 - 1 417 7262745

View Full Product Details