Gene Transcripts

Gene Symbol
RAMP1
Entrez Gene ID
10267
Gene Name
receptor activity modifying protein 1
Gene Aliases
Chromosome Location
Chr. 2: 237858714 - 237912117 on Build GRCh38
UniGene ID
Hs.471783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308353.1 NP_001295282.1 4 866
NM_005855.3 NP_005846.1 3 805
XM_017003152.1 XP_016858641.1 3 727
XM_017003153.1 XP_016858642.1 3 1035
XM_017003154.1 XP_016858643.1 3 812
XM_017003156.1 XP_016858645.1 3 753
GenBank mRNA AJ001014.1 - 3 705
BC000548.2 - 3 697 3163522

View Full Product Details