Gene Transcripts

Gene Symbol
LINC01587
Entrez Gene ID
10141
Gene Name
long intergenic non-protein coding RNA 1587
Gene Aliases
C4orf6, aC1
Chromosome Location
Chr. 4: 5524569 - 5527801 on Build GRCh38
UniGene ID
Hs.177972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_126517.1 1 258
NR_126518.1 1 258
GenBank mRNA BC117444.1 - 1 212
BC117446.1 - 1 212 40125997
BX094519.1 - 1 76
D82070.1 - 1 258

View Full Product Details