Gene Transcripts
- Gene Symbol
- ASPSCR1
- Entrez Gene ID
- 79058
- Gene Name
- ASPSCR1, UBX domain containing tether for SLC2A4
- Gene Aliases
- ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
- Chromosome Location
- Chr. 17: 81977550 - 82018929 on Build GRCh38
- UniGene ID
- Hs.298351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024083.3 |
NP_076988.1
|
12, 13 |
1446 |
|
NR_045351.1 |
|
11, 12 |
1390 |
|
XM_011523601.2 |
XP_011521903.1
|
12, 13 |
1446 |
|
XM_011523602.2 |
XP_011521904.1
|
12, 13 |
1446 |
|
XM_017025036.1 |
XP_016880525.1
|
12, 13 |
1521 |
|
XM_017025037.1 |
XP_016880526.1
|
12, 13 |
1521 |
|
XM_017025038.1 |
XP_016880527.1
|
13, 14 |
1532 |
|
XM_017025039.1 |
XP_016880528.1
|
13, 14 |
1449 |
|
XM_017025040.1 |
XP_016880529.1
|
12, 13 |
1386 |
|
XM_017025041.1 |
XP_016880530.1
|
11, 12 |
1389 |
|
XM_017025042.1 |
XP_016880531.1
|
12, 13 |
1521 |
|
XM_017025043.1 |
XP_016880532.1
|
11, 12 |
1389 |
|
XM_017025044.1 |
XP_016880533.1
|
13, 14 |
1457 |
|
GenBank mRNA |
AF324219.2 |
- |
12, 13 |
1371 |
|
AK057851.1 |
- |
11, 12 |
1373 |
|
AK090682.1 |
- |
5, 6 |
2811 |
|
AK126967.1 |
- |
5, 6 |
5106 |
|
AK127159.1 |
- |
5, 6 |
2018 |
|
AK226086.1 |
- |
12, 13 |
1319 |
|
AK290624.1 |
- |
12, 13 |
1362 |
|
BC006152.1 |
- |
13, 14 |
1521 |
3357489 |
BC018722.1 |
- |
12, 13 |
1351 |
4640076 |
View Full Product Details