Gene Transcripts

Gene Symbol
TMEM8C
Entrez Gene ID
389827
Gene Name
transmembrane protein 8C
Gene Aliases
MYOMAKER, TMEM226
Chromosome Location
Chr. 9: 133514586 - 133524946 on Build GRCh38
UniGene ID
Hs.512467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080483.2 NP_001073952.1 3 367

View Full Product Details