Gene Transcripts

Gene Symbol
HS6ST1
Entrez Gene ID
9394
Gene Name
heparan sulfate 6-O-sulfotransferase 1
Gene Aliases
HH15, HS6ST
Chromosome Location
Chr. 2: 128265480 - 128318597 on Build GRCh38
UniGene ID
Hs.512841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004807.2 NP_004798.3 2 2229
GenBank mRNA AK058121.1 - 1 53
BC001196.1 - 1 1478

View Full Product Details