Gene Transcripts

Gene Symbol
PLIN3
Entrez Gene ID
10226
Gene Name
perilipin 3
Gene Aliases
M6PRBP1, PP17, TIP47
Chromosome Location
Chr. 19: 4838334 - 4867768 on Build GRCh38
UniGene ID
Hs.140452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164189.1 NP_001157661.1 8 1702
NM_001164194.1 NP_001157666.1 8 1669
NM_005817.4 NP_005808.3 8 1705
GenBank mRNA AF051315.1 - 4 993
AF055574.1 - 8 1581
AF057140.1 - 8 1602
AK122671.1 - 8 1556
AK225045.1 - 8 1571
AK225400.1 - 8 1586
AK226088.1 - 8 1583
AK291104.1 - 8 1662
BC001590.2 - 8 1578 2987965
BC005818.2 - 8 1594 3833411
BC007566.1 - 8 1594 3028104
BC019278.1 - 8 1594 2905275

View Full Product Details