Gene Transcripts

Gene Symbol
POP7
Entrez Gene ID
10248
Gene Name
POP7 homolog, ribonuclease P/MRP subunit
Gene Aliases
0610037N12Rik, RPP2, RPP20
Chromosome Location
Chr. 7: 100706053 - 100707500 on Build GRCh38
UniGene ID
Hs.416994
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005837.2 NP_005828.2 2 365
GenBank mRNA BC001430.2 - 2 275
U94316.1 - 2 271

View Full Product Details