Gene Transcripts

Gene Symbol
ANKRD36
Entrez Gene ID
375248
Gene Name
ankyrin repeat domain 36
Gene Aliases
UNQ2430
Chromosome Location
Chr. 2: 97113186 - 97264521 on Build GRCh38
UniGene ID
Hs.541894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164315.1 NP_001157787.1 58 3678
XM_006712514.3 XP_006712577.1 58 3988
XM_011511130.1 XP_011509432.1 58 3988
XM_011511131.1 XP_011509433.1 58 3988
XM_017004009.1 XP_016859498.1 58 3988
XM_017004010.1 XP_016859499.1 58 3988
XM_017004011.1 XP_016859500.1 56 3886
XM_017004012.1 XP_016859501.1 56 3886
XM_017004013.1 XP_016859502.1 56 3886
XM_017004014.1 XP_016859503.1 56 3886
XM_017004015.1 XP_016859504.1 56 3886
XM_017004016.1 XP_016859505.1 56 3886
XM_017004017.1 XP_016859506.1 56 3886
XM_017004018.1 XP_016859507.1 56 3886
XM_017004019.1 XP_016859508.1 56 3886
XM_017004020.1 XP_016859509.1 56 3886
XM_017004021.1 XP_016859510.1 56 3886
XM_017004022.1 XP_016859511.1 56 3886
XM_017004023.1 XP_016859512.1 58 3988
XM_017004024.1 XP_016859513.1 57 3850
XM_017004025.1 XP_016859514.1 54 3784
XM_017004026.1 XP_016859515.1 54 3784
XM_017004027.1 XP_016859516.1 54 3784
XM_017004028.1 XP_016859517.1 54 3784
XM_017004029.1 XP_016859518.1 54 3784
XM_017004030.1 XP_016859519.1 54 3784
XM_017004032.1 XP_016859521.1 54 3784
XM_017004033.1 XP_016859522.1 54 3784
XM_017004034.1 XP_016859523.1 54 3784
XM_017004035.1 XP_016859524.1 54 3784
XM_017004036.1 XP_016859525.1 54 3784
XM_017004038.1 XP_016859527.1 54 3784
XM_017004039.1 XP_016859528.1 52 3682
XM_017004040.1 XP_016859529.1 52 3682
XM_017004041.1 XP_016859530.1 52 3682
XM_017004042.1 XP_016859531.1 52 3682
XM_017004044.1 XP_016859533.1 50 3580
XM_017004045.1 XP_016859534.1 50 3580
XM_017004046.1 XP_016859535.1 56 3743
XM_017004047.1 XP_016859536.1 55 3675
XM_017004048.1 XP_016859537.1 54 3602
XM_017004049.1 XP_016859538.1 56 3777
XM_017004050.1 XP_016859539.1 41 2181
XM_017004051.1 XP_016859540.1 39 2395

View Full Product Details