Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277174.1 | NP_001264103.1 | 4 | 1240 | |
NM_005864.3 | NP_005855.1 | 4 | 1747 | ||
NM_032459.2 | NP_115835.1 | 3 | 1468 | ||
XM_005267256.1 | XP_005267313.1 | 4 | 1328 | ||
GenBank mRNA | AB001466.1 | - | 4 | 1747 | |
AB001467.1 | - | 3 | 1468 | ||
AK295933.1 | - | 4 | 818 | ||
AK314221.1 | - | 4 | 1309 | ||
BC034246.1 | - | 3 | 1016 | 4214788 | |
BC046403.1 | - | 3 | 1587 |