Gene Transcripts
- Gene Symbol
- Aup1
- Entrez Gene ID
- 11993
- Gene Name
- ancient ubiquitous protein 1
- Gene Aliases
- AA589454
- Chromosome Location
- Chr. 6: 83054521 - 83057682 on Build GRCm38
- UniGene ID
- Mm.386752
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301649.1 |
NP_001288578.1
|
3, 4 |
627 |
|
NM_007517.4 |
NP_031543.2
|
4, 5 |
778 |
|
GenBank mRNA |
AK154707.1 |
- |
4, 5 |
647 |
|
AK158035.1 |
- |
4, 5 |
569 |
|
AK167335.1 |
- |
4, 5 |
568 |
|
AK171011.1 |
- |
4, 5 |
581 |
|
AK171184.1 |
- |
4, 5 |
574 |
|
AK171202.1 |
- |
4, 5 |
588 |
|
AK171249.1 |
- |
4, 5 |
568 |
|
AK171541.1 |
- |
4, 5 |
568 |
|
AK195286.1 |
- |
3, 4 |
229 |
|
BC012699.1 |
- |
3, 4 |
411 |
3986517 |
BC016485.1 |
- |
4, 5 |
571 |
3962982 |
U41736.1 |
- |
4, 5 |
576 |
|
View Full Product Details