Gene Transcripts

Gene Symbol
Nog
Entrez Gene ID
18121
Gene Name
noggin
Gene Aliases
Chromosome Location
Chr. 11: 89300638 - 89302559 on Build GRCm38
UniGene ID
Mm.135266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008711.2 NP_032737.1 1 719
GenBank mRNA AK146488.1 - 1 722
BC132527.1 - 1 245 40130804
BC138148.1 - 1 245 8861167

View Full Product Details