Gene Transcripts

Gene Symbol
Olfr26
Entrez Gene ID
18324
Gene Name
olfactory receptor 26
Gene Aliases
MOR171-9, MTPCR09
Chromosome Location
Chr. 9: 38855060 - 38855990 on Build GRCm38
UniGene ID
Mm.246531
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146783.2 NP_666994.2 1 275
GenBank mRNA BC130238.1 - 1 275

View Full Product Details