Gene Transcripts

Gene Symbol
Prss3
Entrez Gene ID
22073
Gene Name
protease, serine 3
Gene Aliases
MTG, TRY4, Tb, Try3
Chromosome Location
Chr. 6: 41373759 - 41377613 on Build GRCm38
UniGene ID
Mm.425318
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011645.2 NP_035775.1 5 633
GenBank mRNA AK131862.1 - 5 634
BC098498.1 - 5 616

View Full Product Details