Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163626.1 | NP_001157098.1 | 1, 2 | 467 | |
NM_172204.4 | NP_757341.3 | 1, 2 | 467 | ||
GenBank mRNA | AF539798.1 | - | 1, 2 | 186 | |
AK087263.1 | - | 1, 2 | 393 | ||
AY255770.1 | - | 1, 2 | 186 | ||
BC047532.1 | - | 1, 2 | 458 | 4988389 |