Gene Transcripts

Gene Symbol
Hist3h2bb-ps
Entrez Gene ID
382522
Gene Name
histone cluster 3, H2bb, pseudogene
Gene Aliases
4930534G10Rik, 4933432H21, B2Hist3h2bb, Hist3h2bb, R75370
Chromosome Location
Chr. 11: 58954048 - 58954512 on Build GRCm38
UniGene ID
Mm.377898
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206882.1 NP_996765.1 1 41
GenBank mRNA BC107301.1 - 1 40
BC107302.1 - 1 41 40053184

View Full Product Details