Gene Transcripts

Gene Symbol
Asb9
Entrez Gene ID
69299
Gene Name
ankyrin repeat and SOCS box-containing 9
Gene Aliases
1700011M07Rik
Chromosome Location
Chr. X: 164497903 - 164539752 on Build GRCm38
UniGene ID
Mm.79031
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027027.2 NP_081303.1 929
GenBank mRNA AF398970.1 - 7 861
AK005878.1 - 930
BC061038.1 - 921 6703903

View Full Product Details