Gene Transcripts

Gene Symbol
OCLM
Entrez Gene ID
10896
Gene Name
oculomedin
Gene Aliases
TISR
Chromosome Location
Chr. 1: 186400572 - 186401455 on Build GRCh38
UniGene ID
Hs.679230
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022375.3 NP_071770.1 1 702
GenBank mRNA AF142063.1 - 1 817
BC069096.1 - 1 719 7216910

View Full Product Details