Gene Transcripts

Gene Symbol
GADD45G
Entrez Gene ID
10912
Gene Name
growth arrest and DNA damage inducible gamma
Gene Aliases
CR6, DDIT2, GADD45gamma, GRP17
Chromosome Location
Chr. 9: 89605009 - 89606555 on Build GRCh38
UniGene ID
Hs.9701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006705.3 NP_006696.1 4 1033
XM_011518163.2 XP_011516465.1 3 934
GenBank mRNA AF079806.1 - 4 1021
AF087883.1 - 4 1015
BC000465.2 - 4 1014 2822765
BC019325.1 - 4 1014 2823131
D83023.1 - 4 1006

View Full Product Details