Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193270.2 | NP_001180199.1 | 3 | 311 | |
NM_001282174.1 | NP_001269103.1 | 3 | 326 | ||
NM_006800.3 | NP_006791.2 | 2 | 296 | ||
NM_078628.1 | NP_523352.1 | 3 | 326 | ||
NM_078629.3 | NP_523353.2 | 3 | 326 | ||
XM_005274440.2 | XP_005274497.1 | 3 | 203 | ||
XM_011545442.2 | XP_011543744.1 | 3 | 280 | ||
GenBank mRNA | AF117065.1 | - | 3 | 326 | |
AK025642.1 | - | 2 | 270 | ||
AK289780.1 | - | 3 | 248 | ||
AK294255.1 | - | 3 | 196 | ||
AK300814.1 | - | 3 | 284 | ||
AK304419.1 | - | 2 | 124 | ||
BC031210.1 | - | 3 | 259 | 5313214 | |
BI091178.1 | - | 3 | 268 | ||
CT002798.1 | - | 2 | 360 | ||
DA172776.1 | - | 3 | 226 | ||
DA788603.1 | - | 2 | 298 | ||
DA961602.1 | - | 1 | 397 | ||
DC395802.1 | - | 3 | 284 |