Gene Transcripts
- Gene Symbol
- RAB35
- Entrez Gene ID
- 11021
- Gene Name
- RAB35, member RAS oncogene family
- Gene Aliases
- H-ray, RAB1C, RAY
- Chromosome Location
- Chr. 12: 120095094 - 120116839 on Build GRCh38
- UniGene ID
- Hs.524788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006861.6 |
NP_006852.1
|
5, 6 |
669 |
|
XM_011537779.1 |
XP_011536081.1
|
5, 6 |
606 |
|
GenBank mRNA |
AF498960.1 |
- |
5, 6 |
480 |
|
AK125543.1 |
- |
6, 7 |
1444 |
|
AK312538.1 |
- |
5, 6 |
579 |
|
BC015931.2 |
- |
5, 6 |
584 |
3907209 |
BM467132.1 |
- |
5, 6 |
651 |
|
BQ233709.1 |
- |
5, 6 |
625 |
|
BT020024.1 |
- |
5, 6 |
480 |
|
CR536486.1 |
- |
5, 6 |
480 |
|
CR541683.1 |
- |
5, 6 |
480 |
|
X79781.1 |
- |
5, 6 |
561 |
|
View Full Product Details